Product Details

SNP ID
rs12092512
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:54032310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCATAGATACTCAGCTTCTGC[A/C]AAAGAAAACCAATGTACATTAGTAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 Intron NP_001291972.1
NM_001305049.1 Intron NP_001291978.1
NM_001305050.1 Intron NP_001291979.1
NM_001305051.1 Intron NP_001291980.1
NM_001305052.1 Intron NP_001291981.1
NM_001305066.1 Intron NP_001291995.1
NM_004872.4 Intron NP_004863.2
XM_006711051.1 Intron XP_006711114.1

View Full Product Details