Product Details

SNP ID
rs10518693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40407823 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCACACAGTTTTCTGGTAAATGAAG[C/T]CTCTCTAAGAATGCAGCCCCTCTCT
Phenotype
MIM: 607036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IVD PubMed Links

Gene Details

Gene
IVD
Gene Name
isovaleryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159508.1 Intron NP_001152980.1
NM_002225.3 Intron NP_002216.2
XM_005254350.2 Intron XP_005254407.1
XM_006720492.2 Intron XP_006720555.1
XM_006720495.2 Intron XP_006720558.1
XM_017022149.1 Intron XP_016877638.1
XM_017022150.1 Intron XP_016877639.1
XM_017022151.1 Intron XP_016877640.1
XM_017022152.1 Intron XP_016877641.1
XM_017022153.1 Intron XP_016877642.1
XM_017022154.1 Intron XP_016877643.1
XM_017022155.1 Intron XP_016877644.1
XM_017022156.1 Intron XP_016877645.1
XM_017022157.1 Intron XP_016877646.1
XM_017022158.1 Intron XP_016877647.1

View Full Product Details