Product Details
- SNP ID
-
rs59160527
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:63588878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCTTGTTTCCTATGCACAAATTC[A/T]TTTGGCGGGGGGGTTGTCAGCCCTC
- Phenotype
-
MIM: 604445
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SERPINB13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs562269359] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SERPINB13
- Gene Name
- serpin family B member 13
View Full Product Details