Product Details

SNP ID
rs7147309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24137243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGGAGCTAGGGAGTAAAGGCC[A/G]AGAGAAGAGTCTGGAGGCTGTGAGA
Phenotype
MIM: 612028 MIM: 600654 MIM: 602161 MIM: 612487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMC9 PubMed Links

Gene Details

Gene
EMC9
Gene Name
ER membrane protein complex subunit 9
There are no transcripts associated with this gene.

Gene
FITM1
Gene Name
fat storage inducing transmembrane protein 1
There are no transcripts associated with this gene.

Gene
MIR7703
Gene Name
microRNA 7703
There are no transcripts associated with this gene.

Gene
PSME1
Gene Name
proteasome activator subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281528.1 Intron NP_001268457.1
NM_001281529.1 Intron NP_001268458.1
NM_006263.3 Intron NP_006254.1
NM_176783.2 Intron NP_788955.1
Gene
PSME2
Gene Name
proteasome activator subunit 2
There are no transcripts associated with this gene.

Gene
RNF31
Gene Name
ring finger protein 31
There are no transcripts associated with this gene.

View Full Product Details