Product Details
- SNP ID
-
rs11554674
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
55
- Location
-
Chr.1:33324895 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTCCTTGAGCATGCTGATGCGGGC[A/G]TAGATCTTCAGGGCGGGCCCCAGCT
- Phenotype
-
MIM: 602979
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
A3GALT2
PubMed Links
Gene Details
- Gene
- A3GALT2
- Gene Name
- alpha 1,3-galactosyltransferase 2
There are no transcripts associated with this gene.
- Gene
- MIR3605
- Gene Name
- microRNA 3605
There are no transcripts associated with this gene.
- Gene
- PHC2
- Gene Name
- polyhomeotic homolog 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004427.3 |
2978 |
Silent Mutation |
TAC,TAT |
Y314Y |
NP_004418.2 |
NM_198040.2 |
2978 |
Silent Mutation |
TAC,TAT |
Y849Y |
NP_932157.1 |
XM_005270570.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y314Y |
XP_005270627.1 |
XM_011540876.2 |
2978 |
Intron |
|
|
XP_011539178.1 |
XM_011540877.2 |
2978 |
Intron |
|
|
XP_011539179.1 |
XM_011540878.2 |
2978 |
Intron |
|
|
XP_011539180.1 |
XM_017000513.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y872Y |
XP_016856002.1 |
XM_017000514.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y872Y |
XP_016856003.1 |
XM_017000515.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y872Y |
XP_016856004.1 |
XM_017000516.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y850Y |
XP_016856005.1 |
XM_017000517.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y850Y |
XP_016856006.1 |
XM_017000518.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y850Y |
XP_016856007.1 |
XM_017000519.1 |
2978 |
UTR 3 |
|
|
XP_016856008.1 |
XM_017000520.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y821Y |
XP_016856009.1 |
XM_017000521.1 |
2978 |
Silent Mutation |
TAC,TAT |
Y821Y |
XP_016856010.1 |
XM_017000522.1 |
2978 |
Intron |
|
|
XP_016856011.1 |
View Full Product Details