Product Details

SNP ID
rs41301281
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:117112471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACCCACCTACATGGGACAAAGAG[A/G]AAAGGACAAAAATCAACCATTAGCG
Phenotype
MIM: 609318 MIM: 604718
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM45 PubMed Links

Gene Details

Gene
TRIM45
Gene Name
tripartite motif containing 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145635.1 Intron NP_001139107.1
NM_025188.3 Intron NP_079464.2
XM_011542199.2 Intron XP_011540501.1
Gene
TTF2
Gene Name
transcription termination factor 2
There are no transcripts associated with this gene.

View Full Product Details