Product Details

SNP ID
rs34586111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24291155 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCTCACCGTCCACATCCCGAAG[G/T]CCATAGCGTCGAGCAAGGTCACAGG
Phenotype
MIM: 610410
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DHRS1 PubMed Links

Gene Details

Gene
DHRS1
Gene Name
dehydrogenase/reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136050.2 1058 Silent Mutation GGA,GGC G263G NP_001129522.1
NM_138452.2 1058 Silent Mutation GGA,GGC G263G NP_612461.1
Gene
NOP9
Gene Name
NOP9 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details