Product Details

SNP ID
rs35670478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50242479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGGCTTCTGAGCACTCACTGG[C/T]ACACTGTTGCAACTGAGGGAGCGCA
Phenotype
MIM: 142704 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDC PubMed Links

Gene Details

Gene
HDC
Gene Name
histidine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306146.1 1981 Silent Mutation GTA,GTG V557V NP_001293075.1
NM_002112.3 1981 Silent Mutation GTA,GTG V590V NP_002103.2
XM_017022094.1 1981 Silent Mutation GTA,GTG V625V XP_016877583.1
XM_017022095.1 1981 Silent Mutation GTA,GTG V592V XP_016877584.1
XM_017022096.1 1981 Silent Mutation GTA,GTG V549V XP_016877585.1
XM_017022097.1 1981 Silent Mutation GTA,GTG V546V XP_016877586.1
XM_017022098.1 1981 Silent Mutation GTA,GTG V481V XP_016877587.1
XM_017022099.1 1981 Intron XP_016877588.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
There are no transcripts associated with this gene.

View Full Product Details