Product Details

SNP ID
rs62623668
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCGCGCCAGACGCCACACAAGC[C/T]TTTTCTGCTCTTCCGCGACGAGACT
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 441 Intron NP_079113.2
XM_011522046.2 441 Intron XP_011520348.1
XM_011522047.2 441 Intron XP_011520349.1
XM_011522048.1 441 Intron XP_011520350.1
XM_011522049.2 441 Intron XP_011520351.1
XM_011522051.2 441 Intron XP_011520353.1
XM_011522052.2 441 Intron XP_011520354.1
XM_011522053.1 441 Intron XP_011520355.1
XM_011522056.2 441 Intron XP_011520358.2
XM_011522058.2 441 Intron XP_011520360.1
XM_011522059.1 441 Intron XP_011520361.1
XM_011522060.1 441 Intron XP_011520362.1
XM_011522061.1 441 Intron XP_011520363.1
XM_011522062.1 441 Intron XP_011520364.1
XM_011522063.1 441 Intron XP_011520365.1
XM_011522064.1 441 Intron XP_011520366.1
XM_011522069.2 441 Intron XP_011520371.1
XM_011522070.1 441 Intron XP_011520372.1
XM_017022587.1 441 Intron XP_016878076.1
XM_017022588.1 441 Intron XP_016878077.1
XM_017022589.1 441 Intron XP_016878078.1
XM_017022590.1 441 Intron XP_016878079.1
XM_017022591.1 441 Intron XP_016878080.1
XM_017022592.1 441 Intron XP_016878081.1
XM_017022593.1 441 Intron XP_016878082.1
XM_017022594.1 441 Intron XP_016878083.1
XM_017022595.1 441 Intron XP_016878084.1
XM_017022596.1 441 Intron XP_016878085.1
XM_017022597.1 441 Intron XP_016878086.1
XM_017022598.1 441 Intron XP_016878087.1
XM_017022599.1 441 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 441 Missense Mutation CCT,CTT P70L NP_001153101.1
NM_003645.3 441 Missense Mutation CCT,CTT P70L NP_003636.2

View Full Product Details