Product Details

SNP ID
rs9804371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:114285910 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGCATGCCGCATGACCATGGCTT[A/G]ACAGTGGGTGTTGCTGTGATCCCCG
Phenotype
MIM: 612420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFAP1L2 PubMed Links

Gene Details

Gene
AFAP1L2
Gene Name
actin filament associated protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001936.2 Intron NP_001001936.1
NM_001287824.1 Intron NP_001274753.1
NM_032550.3 Intron NP_115939.1
XM_005270233.4 Intron XP_005270290.1
XM_005270237.4 Intron XP_005270294.1
XM_005270241.4 Intron XP_005270298.1
XM_005270242.4 Intron XP_005270299.1
XM_011540286.2 Intron XP_011538588.1
XM_011540289.1 Intron XP_011538591.1
XM_011540290.1 Intron XP_011538592.1
XM_017016792.1 Intron XP_016872281.1
XM_017016793.1 Intron XP_016872282.1
XM_017016794.1 Intron XP_016872283.1
XM_017016795.1 Intron XP_016872284.1
XM_017016796.1 Intron XP_016872285.1
XM_017016797.1 Intron XP_016872286.1
XM_017016798.1 Intron XP_016872287.1
XM_017016799.1 Intron XP_016872288.1
XM_017016800.1 Intron XP_016872289.1
XM_017016801.1 Intron XP_016872290.1
XM_017016802.1 Intron XP_016872291.1
XM_017016803.1 Intron XP_016872292.1
XM_017016804.1 Intron XP_016872293.1
XM_017016805.1 Intron XP_016872294.1
XM_017016806.1 Intron XP_016872295.1
XM_017016807.1 Intron XP_016872296.1
XM_017016808.1 Intron XP_016872297.1
XM_017016809.1 Intron XP_016872298.1
XM_017016810.1 Intron XP_016872299.1
XM_017016811.1 Intron XP_016872300.1
XM_017016812.1 Intron XP_016872301.1
XM_017016813.1 Intron XP_016872302.1
XM_017016814.1 Intron XP_016872303.1
XM_017016815.1 Intron XP_016872304.1
XM_017016816.1 Intron XP_016872305.1
XM_017016817.1 Intron XP_016872306.1
XM_017016818.1 Intron XP_016872307.1
XM_017016819.1 Intron XP_016872308.1
XM_017016820.1 Intron XP_016872309.1
XM_017016821.1 Intron XP_016872310.1
XM_017016822.1 Intron XP_016872311.1
XM_017016823.1 Intron XP_016872312.1
XM_017016824.1 Intron XP_016872313.1
XM_017016825.1 Intron XP_016872314.1
XM_017016826.1 Intron XP_016872315.1
XM_017016827.1 Intron XP_016872316.1
XM_017016828.1 Intron XP_016872317.1
XM_017016829.1 Intron XP_016872318.1
XM_017016830.1 Intron XP_016872319.1
XM_017016831.1 Intron XP_016872320.1
XM_017016832.1 Intron XP_016872321.1
Gene
VWA2
Gene Name
von Willebrand factor A domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272046.1 Intron NP_001258975.1
NM_001320804.1 Intron NP_001307733.1
XM_011539754.2 Intron XP_011538056.1
XM_011539757.2 Intron XP_011538059.1
XM_017016177.1 Intron XP_016871666.1
XM_017016178.1 Intron XP_016871667.1
XM_017016179.1 Intron XP_016871668.1
XM_017016180.1 Intron XP_016871669.1
XM_017016181.1 Intron XP_016871670.1

View Full Product Details