Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001272046.1 | 293 | Missense Mutation | ACC,GCC | T9A | NP_001258975.1 |
NM_001320804.1 | 293 | Missense Mutation | ACC,GCC | T9A | NP_001307733.1 |
XM_011539754.2 | 293 | UTR 5 | XP_011538056.1 | ||
XM_011539757.2 | 293 | Missense Mutation | ACC,GCC | T9A | XP_011538059.1 |
XM_017016177.1 | 293 | Missense Mutation | ACC,GCC | T19A | XP_016871666.1 |
XM_017016178.1 | 293 | Missense Mutation | ACC,GCC | T19A | XP_016871667.1 |
XM_017016179.1 | 293 | Missense Mutation | ACC,GCC | T15A | XP_016871668.1 |
XM_017016180.1 | 293 | Intron | XP_016871669.1 | ||
XM_017016181.1 | 293 | Missense Mutation | ACC,GCC | T19A | XP_016871670.1 |