Product Details

SNP ID
rs62641685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:13322914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCACCGCAGCCATCTTGGCCAG[C/G]ACCGGGAGGTTGTGAATTACAAACG
Phenotype
MIM: 600902
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SEPHS1 PubMed Links

Gene Details

Gene
SEPHS1
Gene Name
selenophosphate synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195602.1 1251 Intron NP_001182531.1
NM_001195604.1 1251 Intron NP_001182533.1
NM_012247.4 1251 Intron NP_036379.2
XM_017015942.1 1251 Silent Mutation GTC,GTG V295V XP_016871431.1
XM_017015943.1 1251 Silent Mutation GTC,GTG V295V XP_016871432.1
XM_017015944.1 1251 Silent Mutation GTC,GTG V293V XP_016871433.1
XM_017015945.1 1251 Intron XP_016871434.1

View Full Product Details