Product Details

SNP ID
rs35722563
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17812299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGTGGTCTATAAACGTGAACCA[C/G]CTGTGGAGAGGGTAATAGAATTTGC
Phenotype
MIM: 606280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCAF16 PubMed Links

Gene Details

Gene
DCAF16
Gene Name
DDB1 and CUL4 associated factor 16
There are no transcripts associated with this gene.

Gene
NCAPG
Gene Name
non-SMC condensin I complex subunit G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022346.4 455 Missense Mutation CCT,GCT P64A NP_071741.2
XM_011513876.2 455 UTR 5 XP_011512178.1
XM_017008543.1 455 Missense Mutation CCT,GCT P64A XP_016864032.1
XM_017008544.1 455 UTR 5 XP_016864033.1

View Full Product Details