Product Details

SNP ID
rs116760479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23580094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGCAGCAGGCCATGGGTTACCA[C/T]GGCCAGACCCAGCAGCAGCAGGGGC
Phenotype
MIM: 146770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRICH1 PubMed Links

Gene Details

Gene
DRICH1
Gene Name
aspartate rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016449.3 455 Intron NP_057533.2
XM_011530206.1 455 Intron XP_011528508.1
XM_011530207.2 455 Intron XP_011528509.1
XM_011530208.2 455 Intron XP_011528510.1
XM_011530209.2 455 Intron XP_011528511.1
XM_011530210.2 455 Intron XP_011528512.1
XM_017028813.1 455 Intron XP_016884302.1
Gene
IGLL1
Gene Name
immunoglobulin lambda like polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020070.3 455 Missense Mutation ATG,GTG M33V NP_064455.1
NM_152855.2 455 Missense Mutation ATG,GTG M33V NP_690594.1
XM_011530169.2 455 Missense Mutation ATG,GTG M33V XP_011528471.1

View Full Product Details