Product Details

SNP ID
rs116013292
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:158181417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTATGCTGGCTGCTCTCCCGG[C/T]CACTTGCTACACGCCTCCAATCTTC
Phenotype
MIM: 188410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD1D PubMed Links

Gene Details

Gene
CD1D
Gene Name
CD1d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319145.1 Intron NP_001306074.1
NM_001766.3 Intron NP_001757.1
XM_005245583.1 Intron XP_005245640.1
XM_006711621.1 Intron XP_006711684.1
XM_011510125.1 Intron XP_011508427.1
XM_011510127.1 Intron XP_011508429.1

View Full Product Details