Product Details

SNP ID
rs4647037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60945141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTGAGCGCCGTTACTAGAAGG[C/G]CTCCAGGATGGAGGCCGACCTGACT
Phenotype
MIM: 609412 MIM: 609653
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ERCC8 PubMed Links

Gene Details

Gene
ERCC8
Gene Name
ERCC excision repair 8, CSA ubiquitin ligase complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000082.3 Intron NP_000073.1
NM_001007233.2 Intron NP_001007234.1
NM_001007234.2 Intron NP_001007235.1
NM_001290285.1 Intron NP_001277214.1
Gene
NDUFAF2
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174889.4 Intron NP_777549.1

View Full Product Details