Product Details

SNP ID
rs143288784
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:192809069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGGGGCTCCAGCGGGAGAACG[A/G]TAATGCAAAGTGCTATGTTCTTGGC
Phenotype
MIM: 600861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RGS2 PubMed Links

Gene Details

Gene
RGS2
Gene Name
regulator of G-protein signaling 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002923.3 31 UTR 5 NP_002914.1

View Full Product Details