Product Details

SNP ID
rs3213467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152829796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCAATACACAAGCACAATAATA[C/T]ATCATACAGTCCATATTTACATAAC
Phenotype
MIM: 300006 MIM: 300275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CETN2 PubMed Links

Gene Details

Gene
CETN2
Gene Name
centrin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004344.1 Intron NP_004335.1
Gene
NSDHL
Gene Name
NAD(P) dependent steroid dehydrogenase-like
There are no transcripts associated with this gene.

View Full Product Details