Product Details

SNP ID
rs114724928
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2464071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATTCTTCCTCCATCCAAGACT[A/T]TGAGTTATCCCTGAAGCAGGGTAGA
Phenotype
MIM: 182282
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
SNORD119 PubMed Links

Gene Details

Gene
SNORD119
Gene Name
small nucleolar RNA, C/D box 119
There are no transcripts associated with this gene.

Gene
SNRPB
Gene Name
small nuclear ribonucleoprotein polypeptides B and B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003091.3 Intron NP_003082.1
NM_198216.1 Intron NP_937859.1

View Full Product Details