Product Details

SNP ID
rs61729225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2102157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGGGAAACGGGCCGCCCGGAA[G/T]TGGAGGTGCGCGGGCCAGGTAAGGG
Phenotype
MIM: 609370
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STK35 PubMed Links

Gene Details

Gene
STK35
Gene Name
serine/threonine kinase 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080836.3 4123 Missense Mutation AAG,AAT K92N NP_543026.2
XM_011529174.2 4123 Missense Mutation AAG,AAT K188N XP_011527476.1

View Full Product Details