Product Details

SNP ID
rs61730770
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20859336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACGAGTCCGAGAAGGTTGGGGTC[G/T]CCTCTTCCGAGGTGAGCCTGGGGCA
Phenotype
MIM: 600286 MIM: 604202
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PI4KA PubMed Links

Gene Details

Gene
PI4KA
Gene Name
phosphatidylinositol 4-kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_058004.3 333 Intron NP_477352.3
XM_005261634.1 333 Intron XP_005261691.1
XM_005261635.1 333 Intron XP_005261692.1
XM_011530226.1 333 Intron XP_011528528.1
XM_011530228.2 333 Intron XP_011528530.1
XM_017028829.1 333 Intron XP_016884318.1
XM_017028830.1 333 Intron XP_016884319.1
Gene
SNAP29
Gene Name
synaptosome associated protein 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004782.3 333 Missense Mutation GCC,TCC A76S NP_004773.1

View Full Product Details