Product Details

SNP ID
rs113555804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22836571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGGGAGGGACCAGAGGAGAGAG[C/T]GAGAGAGGGAACCAGACCCCAGTTC
Phenotype
MIM: 600754 MIM: 611856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP14 PubMed Links

Gene Details

Gene
MMP14
Gene Name
matrix metallopeptidase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004995.3 39 UTR 5 NP_004986.1
Gene
MRPL52
Gene Name
mitochondrial ribosomal protein L52
There are no transcripts associated with this gene.

View Full Product Details