Product Details

SNP ID
hCV25628026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7589694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGCCATGGGTTGGGAGAAGCCTT[A/C]AGAGGGCGTCCTGAATGCCCTCCCC
Phenotype
MIM: 153634 MIM: 605339 MIM: 604041 MIM: 601297
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD68 PubMed Links

Gene Details

Gene
CD68
Gene Name
CD68 molecule
There are no transcripts associated with this gene.

Gene
FXR2
Gene Name
FMR1 autosomal homolog 2
There are no transcripts associated with this gene.

Gene
LOC100996842
Gene Name
uncharacterized LOC100996842
There are no transcripts associated with this gene.

Gene
MPDU1
Gene Name
mannose-P-dolichol utilization defect 1
There are no transcripts associated with this gene.

Gene
SOX15
Gene Name
SRY-box 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006942.1 477 UTR 5 NP_008873.1

View Full Product Details