Product Details

SNP ID
rs16932266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:65605314 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGCTGAGCTTTCATAACCTTGGT[C/T]GATGCTATTGCTGATGCCAAAGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARMC1 PubMed Links

Gene Details

Gene
ARMC1
Gene Name
armadillo repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286702.1 Intron NP_001273631.1
NM_018120.5 Intron NP_060590.1
XM_005251264.1 Intron XP_005251321.1
XM_017013605.1 Intron XP_016869094.1

View Full Product Details