Product Details

SNP ID
rs55710619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57232641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGGTGAGCTCTACAAGGAGCT[A/G]CAGAAAAGCGAGAAATTAGATGAAC
Phenotype
MIM: 603495 MIM: 604668
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKC PubMed Links

Gene Details

Gene
AURKC
Gene Name
aurora kinase C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015878.1 444 Silent Mutation CTA,CTG L132L NP_001015878.1
NM_001015879.1 444 Silent Mutation CTA,CTG L113L NP_001015879.1
NM_003160.2 444 Silent Mutation CTA,CTG L98L NP_003151.2
Gene
ZNF264
Gene Name
zinc finger protein 264
There are no transcripts associated with this gene.

Gene
ZNF805
Gene Name
zinc finger protein 805
There are no transcripts associated with this gene.

View Full Product Details