Product Details

SNP ID
rs200955684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52053883 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTCAGGCGGGGGCGCCACCCA[C/G]AGTCCTGCATACACCTTGATGCACC
Phenotype
MIM: 602749
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP7 PubMed Links

Gene Details

Gene
DUSP7
Gene Name
dual specificity phosphatase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001947.3 Intron NP_001938.2
Gene
LINC00696
Gene Name
long intergenic non-protein coding RNA 696
There are no transcripts associated with this gene.

View Full Product Details