Product Details

SNP ID
rs80222959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179617497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCACCTGTTAAGAGCCCACAAA[A/T]GCTCAATCACACTTACGCATATTTG
Phenotype
MIM: 601035 MIM: 610327
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPH1 PubMed Links

Gene Details

Gene
HNRNPH1
Gene Name
heterogeneous nuclear ribonucleoprotein H1 (H)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257293.1 Intron NP_001244222.1
NM_005520.2 Intron NP_005511.1
XM_005265895.2 Intron XP_005265952.1
XM_005265896.2 Intron XP_005265953.1
XM_005265901.1 Intron XP_005265958.1
XM_005265902.4 Intron XP_005265959.1
XM_006714862.2 Intron XP_006714925.1
XM_006714863.2 Intron XP_006714926.1
XM_011534542.1 Intron XP_011532844.1
XM_011534544.1 Intron XP_011532846.1
XM_011534545.1 Intron XP_011532847.1
XM_011534546.1 Intron XP_011532848.1
XM_011534547.1 Intron XP_011532849.1
XM_017009414.1 Intron XP_016864903.1
XM_017009415.1 Intron XP_016864904.1
XM_017009416.1 Intron XP_016864905.1
XM_017009417.1 Intron XP_016864906.1
XM_017009418.1 Intron XP_016864907.1
XM_017009419.1 Intron XP_016864908.1
XM_017009420.1 Intron XP_016864909.1
XM_017009421.1 Intron XP_016864910.1
XM_017009422.1 Intron XP_016864911.1
XM_017009423.1 Intron XP_016864912.1
XM_017009424.1 Intron XP_016864913.1
XM_017009425.1 Intron XP_016864914.1
Gene
RUFY1
Gene Name
RUN and FYVE domain containing 1
There are no transcripts associated with this gene.

View Full Product Details