Product Details

SNP ID
rs6770808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184347276 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGACGGAGAAATAGGATGGGGGA[A/G]TAATTCTTAGTCCCATGTTAGAAAG
Phenotype
MIM: 600570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCN2 PubMed Links

Gene Details

Gene
CLCN2
Gene Name
chloride voltage-gated channel 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171087.2 Intron NP_001164558.1
NM_001171088.2 Intron NP_001164559.1
NM_001171089.2 Intron NP_001164560.1
NM_004366.5 Intron NP_004357.3
XM_006713489.1 Intron XP_006713552.1
XM_006713490.2 Intron XP_006713553.1
XM_011512401.1 Intron XP_011510703.1
Gene
FAM131A
Gene Name
family with sequence similarity 131 member A
There are no transcripts associated with this gene.

View Full Product Details