Product Details

SNP ID
rs35555615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39972455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACCTGAAAAAGGAATTTCTCCA[C/T]GCCCAGGAGGAGGTGAAGCGAATCC
Phenotype
MIM: 602707
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMC4 PubMed Links

Gene Details

Gene
PSMC4
Gene Name
proteasome 26S subunit, ATPase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006503.3 420 Silent Mutation CAC,CAT H74H NP_006494.1
NM_153001.2 420 Intron NP_694546.1

View Full Product Details