Product Details

SNP ID
rs79475116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39393528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAAGATTAGAAATCAATTCTTCA[A/G]ACATCCTTTTTTCCTTGTCTGTAGA
Phenotype
MIM: 612914 MIM: 610506
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED29 PubMed Links

Gene Details

Gene
MED29
Gene Name
mediator complex subunit 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317770.1 Intron NP_001304699.1
NM_017592.2 Intron NP_060062.1
Gene
PAF1
Gene Name
PAF1 homolog, Paf1/RNA polymerase II complex component
There are no transcripts associated with this gene.

Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
There are no transcripts associated with this gene.

View Full Product Details