Product Details
- SNP ID
-
rs371913287
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:53191509 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCCTTCCTCTCACCCTCCAGTTCC[A/C]ACTGTCCTCCAAGGAGAAGAGCCTT
- Phenotype
-
MIM: 147559
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ITGB7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80201470] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ITGB7
- Gene Name
- integrin subunit beta 7
- Gene
- ZNF740
- Gene Name
- zinc finger protein 740
View Full Product Details