Product Details

SNP ID
rs3737502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:108443634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACTGCGTCTCCCCGCCCTCTCAC[C/T]CCGCTGCCTCTAGGTTCTGGGAAGA
Phenotype
MIM: 607722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC2 PubMed Links

Gene Details

Gene
EMC2
Gene Name
ER membrane protein complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details