Product Details

SNP ID
rs150933455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:134566368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGGGATCACAAACTTAGATGG[C/T]GTGCCCTTAGAAGAGTAGTGTATCT
Phenotype
MIM: 300296
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00629 PubMed Links

Gene Details

Gene
LINC00629
Gene Name
long intergenic non-protein coding RNA 629
There are no transcripts associated with this gene.

Gene
PLAC1
Gene Name
placenta specific 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316887.1 601 Silent Mutation ACA,ACG T105T NP_001303816.1
NM_001316888.1 601 Silent Mutation ACA,ACG T105T NP_001303817.1
NM_001316889.1 601 Silent Mutation ACA,ACG T105T NP_001303818.1
NM_021796.3 601 Silent Mutation ACA,ACG T105T NP_068568.1
XM_011531257.2 601 Silent Mutation ACA,ACG T105T XP_011529559.1
XM_011531260.2 601 Silent Mutation ACA,ACG T105T XP_011529562.1
XM_017029236.1 601 Silent Mutation ACA,ACG T105T XP_016884725.1

View Full Product Details