Product Details

SNP ID
rs76414810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47498277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGAAGCCTGTTGCCCAGGAGGCC[C/T]CTGCATACCTGCCCGTGCACATCCC
Phenotype
MIM: 615020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELP6 PubMed Links

Gene Details

Gene
ELP6
Gene Name
elongator acetyltransferase complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031703.2 Intron NP_001026873.2
XM_005265241.3 Intron XP_005265298.1
XM_011533851.2 Intron XP_011532153.1
XM_011533852.2 Intron XP_011532154.1
XM_011533853.2 Intron XP_011532155.1
XM_011533854.2 Intron XP_011532156.1
XM_011533856.2 Intron XP_011532158.1
XM_011533857.2 Intron XP_011532159.1
XM_017006669.1 Intron XP_016862158.1
XM_017006670.1 Intron XP_016862159.1

View Full Product Details