Product Details

SNP ID
rs200268742
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29610370 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGGGATCCTGGGCTGCCCAGA[C/G]CGGAGCAGTGAGCGCCGCACCCCCT
Phenotype
MIM: 606114
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POP4 PubMed Links

Gene Details

Gene
POP4
Gene Name
POP4 homolog, ribonuclease P/MRP subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006627.2 Intron NP_006618.1

View Full Product Details