Product Details

SNP ID
rs79943460
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:39847192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATATTCTATTGAAAACAGACCC[A/G]TAGGATAAAGAAAAACATGAGACTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIT1 PubMed Links

Gene Details

Gene
TRIT1
Gene Name
tRNA isopentenyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001312691.1 Intron NP_001299620.1
NM_001312692.1 Intron NP_001299621.1
NM_017646.5 Intron NP_060116.2
XM_005270954.2 Intron XP_005271011.1
XM_006710706.1 Intron XP_006710769.1

View Full Product Details