Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001178000.1 | Intron | NP_001171471.1 | |||
NM_015040.3 | Intron | NP_055855.2 | |||
NM_152671.3 | Intron | NP_689884.1 | |||
XM_011510778.2 | Intron | XP_011509080.1 | |||
XM_011510779.2 | Intron | XP_011509081.1 | |||
XM_011510780.2 | Intron | XP_011509082.1 | |||
XM_011510781.2 | Intron | XP_011509083.1 | |||
XM_011510782.2 | Intron | XP_011509084.1 | |||
XM_011510783.2 | Intron | XP_011509085.1 | |||
XM_011510784.2 | Intron | XP_011509086.1 | |||
XM_011510785.2 | Intron | XP_011509087.1 | |||
XM_011510786.2 | Intron | XP_011509088.1 | |||
XM_011510787.1 | Intron | XP_011509089.1 | |||
XM_011510788.1 | Intron | XP_011509090.1 | |||
XM_011510789.2 | Intron | XP_011509091.1 | |||
XM_011510790.1 | Intron | XP_011509092.1 | |||
XM_011510792.2 | Intron | XP_011509094.1 | |||
XM_017003568.1 | Intron | XP_016859057.1 | |||
XM_017003569.1 | Intron | XP_016859058.1 | |||
XM_017003570.1 | Intron | XP_016859059.1 | |||
XM_017003571.1 | Intron | XP_016859060.1 | |||
XM_017003572.1 | Intron | XP_016859061.1 | |||
XM_017003573.1 | Intron | XP_016859062.1 | |||
XM_017003574.1 | Intron | XP_016859063.1 |