Product Details

SNP ID
rs34630572
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44885368 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACTTCTTGAATCAGGACAACGC[C/T]GTTTCTCACCACACATGGGAGTTCC
Phenotype
MIM: 600585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGM4 PubMed Links

Gene Details

Gene
TGM4
Gene Name
transglutaminase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003241.3 136 Silent Mutation GCC,GCT A21A NP_003232.2
XM_011534042.1 136 Silent Mutation GCC,GCT A66A XP_011532344.1

View Full Product Details