Product Details

SNP ID
rs34227468
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:153540056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGAGGGAACAATCACCTACCT[C/T]CCCAAGACACAGCTCTTTCTTGATC
Phenotype
MIM: 176992 MIM: 114210 MIM: 176991 MIM: 114110
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928034 PubMed Links

Gene Details

Gene
LOC101928034
Gene Name
uncharacterized LOC101928034
There are no transcripts associated with this gene.

Gene
S100A3
Gene Name
S100 calcium binding protein A3
There are no transcripts associated with this gene.

Gene
S100A4
Gene Name
S100 calcium binding protein A4
There are no transcripts associated with this gene.

Gene
S100A5
Gene Name
S100 calcium binding protein A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002962.1 908 Missense Mutation AAG,GAG K46E NP_002953.2
XM_011509860.2 908 Missense Mutation AAG,GAG K58E XP_011508162.1
XM_017002029.1 908 Missense Mutation AAG,GAG K87E XP_016857518.1
XM_017002030.1 908 Missense Mutation AAG,GAG K64E XP_016857519.1
XM_017002031.1 908 Missense Mutation AAG,GAG K46E XP_016857520.1
XM_017002032.1 908 Missense Mutation AAG,GAG K46E XP_016857521.1
Gene
S100A6
Gene Name
S100 calcium binding protein A6
There are no transcripts associated with this gene.

View Full Product Details