Product Details

SNP ID
rs139043100
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:155186125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGGCCACTCAGCTCAGCGGG[C/T]GACGTGCCCCTACAAGTTGGCAGAA
Phenotype
MIM: 158340 MIM: 188062 MIM: 600986
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR92B PubMed Links

Gene Details

Gene
MIR92B
Gene Name
microRNA 92b
There are no transcripts associated with this gene.

Gene
MUC1
Gene Name
mucin 1, cell surface associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018016.2 562 UTR 3 NP_001018016.1
NM_001018017.2 562 UTR 3 NP_001018017.1
NM_001044390.2 562 UTR 3 NP_001037855.1
NM_001044391.2 562 UTR 3 NP_001037856.1
NM_001044392.2 562 UTR 3 NP_001037857.1
NM_001044393.2 562 UTR 3 NP_001037858.1
NM_001204285.1 562 UTR 3 NP_001191214.1
NM_001204286.1 562 UTR 3 NP_001191215.1
NM_001204287.1 562 UTR 3 NP_001191216.1
NM_001204288.1 562 UTR 3 NP_001191217.1
NM_001204289.1 562 UTR 3 NP_001191218.1
NM_001204290.1 562 UTR 3 NP_001191219.1
NM_001204291.1 562 UTR 3 NP_001191220.1
NM_001204292.1 562 UTR 3 NP_001191221.1
NM_001204293.1 562 UTR 3 NP_001191222.1
NM_001204294.1 562 UTR 3 NP_001191223.1
NM_001204295.1 562 UTR 3 NP_001191224.1
NM_001204296.1 562 UTR 3 NP_001191225.1
NM_001204297.1 562 UTR 3 NP_001191226.1
NM_002456.5 562 UTR 3 NP_002447.4
Gene
THBS3
Gene Name
thrombospondin 3
There are no transcripts associated with this gene.

Gene
TRIM46
Gene Name
tripartite motif containing 46
There are no transcripts associated with this gene.

View Full Product Details