Product Details

SNP ID
rs61732829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49153431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGTCCTCACCGCTTTCCTCCT[C/T]GCTGGAGGCACCTCCAGCCTCCTCT
Phenotype
MIM: 142705 MIM: 603144 MIM: 606936
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HRC PubMed Links

Gene Details

Gene
HRC
Gene Name
histidine rich calcium binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002152.2 1994 Missense Mutation AAG,GAG K603E NP_002143.1
XM_017026733.1 1994 Missense Mutation AAG,GAG K603E XP_016882222.1
Gene
PPFIA3
Gene Name
PTPRF interacting protein alpha 3
There are no transcripts associated with this gene.

Gene
TRPM4
Gene Name
transient receptor potential cation channel subfamily M member 4
There are no transcripts associated with this gene.

View Full Product Details