Product Details

SNP ID
rs188194710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48795391 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGAGTGCTGGCGTGACGAGATGC[C/T]ACGGGTCGGTGGATCTGGAGCACAG
Phenotype
MIM: 113530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAT2 PubMed Links

Gene Details

Gene
BCAT2
Gene Name
branched chain amino acid transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164773.1 1751 UTR 3 NP_001158245.1
NM_001190.3 1751 UTR 3 NP_001181.2
NM_001284325.1 1751 UTR 3 NP_001271254.1

View Full Product Details