Product Details

SNP ID
rs6766336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171603117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATGGGCACTATGGCCTCTTTGGT[C/T]CCAACAGAAGGCAGTAGGCTTTCTT
Phenotype
MIM: 602382
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLD1 PubMed Links

Gene Details

Gene
PLD1
Gene Name
phospholipase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130081.2 3268 Silent Mutation GGA,GGG G1024G NP_001123553.1
NM_002662.4 3268 Silent Mutation GGA,GGG G1062G NP_002653.1
XM_005247533.2 3268 Silent Mutation GGA,GGG G1062G XP_005247590.1
XM_005247534.2 3268 Silent Mutation GGA,GGG G1024G XP_005247591.1
XM_011512897.1 3268 Intron XP_011511199.1
XM_011512898.1 3268 Intron XP_011511200.1
XM_011512899.2 3268 Silent Mutation GGA,GGG G645G XP_011511201.1
XM_017006623.1 3268 Intron XP_016862112.1
XM_017006624.1 3268 Silent Mutation GGA,GGG G645G XP_016862113.1

View Full Product Details