Product Details

SNP ID
rs10127895
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:205337567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCCCTGGGTGGAATGCTTCCGC[C/T]GTCTCCAGGACAGTGGGTTGATTCC
Phenotype
MIM: 614503
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHDC8A PubMed Links

Gene Details

Gene
KLHDC8A
Gene Name
kelch domain containing 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271863.1 1429 Silent Mutation ACA,ACG T295T NP_001258792.1
NM_001271864.1 1429 Silent Mutation ACA,ACG T295T NP_001258793.1
NM_001271865.1 1429 Silent Mutation ACA,ACG T295T NP_001258794.1
NM_018203.2 1429 Silent Mutation ACA,ACG T295T NP_060673.1

View Full Product Details