Product Details

SNP ID
hCV25634997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:205338379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGCTTCGAATTTTTTGGAGTAT[A/G]GTGGTAGGCAGGTGGTCTGCAGGGA
Phenotype
MIM: 614503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC8A PubMed Links

Gene Details

Gene
KLHDC8A
Gene Name
kelch domain containing 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271863.1 Intron NP_001258792.1
NM_001271864.1 Intron NP_001258793.1
NM_001271865.1 Intron NP_001258794.1
NM_018203.2 Intron NP_060673.1

View Full Product Details