Product Details

SNP ID
rs35733575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48462310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGCCAAACCCAAGAAGCGCAAC[A/G]AGACGCTGGTCTTCAGCGAGAACGC
Phenotype
MIM: 602488 MIM: 610597 MIM: 603953
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYTH2 PubMed Links

Gene Details

Gene
CYTH2
Gene Name
cytohesin 2
There are no transcripts associated with this gene.

Gene
GRWD1
Gene Name
glutamate rich WD repeat containing 1
There are no transcripts associated with this gene.

Gene
KCNJ14
Gene Name
potassium voltage-gated channel subfamily J member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013348.3 793 Missense Mutation AAG,GAG K196E NP_037480.1
Gene
LOC105372430
Gene Name
uncharacterized LOC105372430
There are no transcripts associated with this gene.

View Full Product Details