Product Details
- SNP ID
-
hCV25635112
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:218454965 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACGGGTAGTCTTCCCCACTTCCG[C/T]GCTAAGTGACTGAGAGTTCTTTTCT
- Phenotype
-
MIM: 193040
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
USP37
PubMed Links
Gene Details
- Gene
- USP37
- Gene Name
- ubiquitin specific peptidase 37
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_020935.2 |
3318 |
Missense Mutation |
ACG,GCG |
T969A |
NP_065986.2 |
- Gene
- VIL1
- Gene Name
- villin 1
There are no transcripts associated with this gene.
View Full Product Details