Product Details

SNP ID
rs73688749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31063362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGAGCCCCAGGCTCACCTGAG[C/T]CCCCGCTCCAGAGAACTCATGTTTT
Phenotype
MIM: 102981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADCYAP1R1 PubMed Links

Gene Details

Gene
ADCYAP1R1
Gene Name
ADCYAP receptor type I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001118.4 Intron NP_001109.2
NM_001199635.1 Intron NP_001186564.1
NM_001199636.1 Intron NP_001186565.1
NM_001199637.1 Intron NP_001186566.1
XM_005249618.4 Intron XP_005249675.1
XM_006715645.2 Intron XP_006715708.1
XM_017011736.1 Intron XP_016867225.1
XM_017011737.1 Intron XP_016867226.1
XM_017011738.1 Intron XP_016867227.1

View Full Product Details