Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171156.1 | Intron | NP_001164627.1 | |||
NM_001171157.1 | Intron | NP_001164628.1 | |||
NM_001171158.1 | Intron | NP_001164629.1 | |||
NM_001171159.1 | Intron | NP_001164630.1 | |||
NM_001171161.1 | Intron | NP_001164632.1 | |||
NM_001322105.1 | Intron | NP_001309034.1 | |||
NM_033130.4 | Intron | NP_149121.2 | |||
XM_005259366.4 | Intron | XP_005259423.1 | |||
XM_005259367.3 | Intron | XP_005259424.1 | |||
XM_011527439.1 | Intron | XP_011525741.1 |