Product Details
- SNP ID
-
rs34152531
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:57471400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCGAAAAGTGGGATTCTCTCTGT[C/T]CTGGTCCTCACCAGTGCTCGTGGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C14orf105
PubMed Links
Gene Details
- Gene
- C14orf105
- Gene Name
- chromosome 14 open reading frame 105
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001283056.1 |
1114 |
Silent Mutation |
AGA,AGG |
R322R |
NP_001269985.1 |
NM_001283057.1 |
1114 |
Silent Mutation |
AGA,AGG |
R281R |
NP_001269986.1 |
NM_001283058.1 |
1114 |
Silent Mutation |
AGA,AGG |
R193R |
NP_001269987.1 |
NM_001283059.1 |
1114 |
Silent Mutation |
AGA,AGG |
R193R |
NP_001269988.1 |
NM_001283060.1 |
1114 |
Intron |
|
|
NP_001269989.1 |
NM_018168.3 |
1114 |
Silent Mutation |
AGA,AGG |
R282R |
NP_060638.2 |
XM_005267806.1 |
1114 |
Intron |
|
|
XP_005267863.1 |
XM_005267810.3 |
1114 |
Silent Mutation |
AGA,AGG |
R203R |
XP_005267867.1 |
XM_005267811.3 |
1114 |
Silent Mutation |
AGA,AGG |
R194R |
XP_005267868.1 |
XM_005267813.1 |
1114 |
Silent Mutation |
AGA,AGG |
R156R |
XP_005267870.1 |
XM_005267814.1 |
1114 |
Silent Mutation |
AGA,AGG |
R155R |
XP_005267871.1 |
XM_006720188.2 |
1114 |
Intron |
|
|
XP_006720251.1 |
XM_006720189.2 |
1114 |
Silent Mutation |
AGA,AGG |
R204R |
XP_006720252.1 |
View Full Product Details